Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhgc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033581.3 | NP_291059.1 | 1 | 1562 | ||
GenBank mRNA | AK086361.1 | - | 1 | 1196 | ||
AK089310.1 | - | 1 | 1609 | |||
AY013811.1 | - | 1 | 1424 | |||
BC040826.1 | - | 1 | 716 | |||
BC059175.1 | - | 1 | 1562 | 5704674 | ||
BC098457.1 | - | 2 | 464 | 30653042 |