Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhgc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033581.3 | NP_291059.1 | 1 | 228 | ||
GenBank mRNA | AK089310.1 | - | 1 | 274 | ||
AY013811.1 | - | 1 | 90 | |||
BC059175.1 | - | 1 | 228 | 5704674 | ||
BC098457.1 | - | 1 | 222 | 30653042 |
Gene Symbol: BC037039 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142308.1 | - | 1 | 343 | ||
BC037039.1 | - | 1 | 1293 |