Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhga4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033587.3 | NP_291065.3 | 1 | 667 | ||
GenBank mRNA | AY013798.1 | - | 1 | 667 | ||
BC049814.1 | - | 1 | 793 | 6508140 |
Gene Symbol: Pcdhgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082842.1 | - | 2 | 3459 |