Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033588.4 | NP_291066.1 | 1 | 362 | ||
GenBank mRNA | AK043161.1 | - | 1 | 477 | ||
AK043491.1 | - | 1 | 439 | |||
AY013799.1 | - | 1 | 362 | |||
BC057404.1 | - | 1 | 462 | 6405582 |