Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033590.3 | NP_291068.2 | 1 | 1012 | ||
GenBank mRNA | AK034910.1 | - | 1 | 1029 | ||
AY013801.1 | - | 1 | 904 | |||
BC077712.1 | - | 1 | 1012 | 6825399 |