Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205396.1 | NP_001192325.1 | 2 | 779 | ||
NM_053118.1 | NP_444348.1 | 2 | 779 | |||
GenBank mRNA | AB025221.1 | - | 1 | 625 | ||
AB025222.1 | - | 1 | 625 | |||
AB099816.1 | - | 1 | 670 | |||
AF218809.1 | - | 2 | 779 | |||
AK137739.1 | - | 2 | 742 |
Set Membership: |
![]() |