Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080819.1 | NP_001074288.1 | 20 | 5173 | ||
GenBank mRNA | AF268912.1 | - | 19 | 4036 | ||
BC082554.1 | - | 4417 | 30622048 |