Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290708.1 | NP_001277637.1 | 11 | 1546 | ||
NM_053123.4 | NP_444353.3 | 11 | 1546 | |||
GenBank mRNA | AF325920.1 | - | 11 | 1553 | ||
AK030741.1 | - | 11 | 1513 | |||
AK032978.1 | - | 11 | 1514 | |||
AK052092.1 | - | 11 | 1502 | |||
BC016071.1 | - | 11 | 1523 | |||
BC057115.1 | - | 11 | 1546 | 6406330 |