Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199351.1 | NP_001186280.1 | 11 | 1167 | ||
NM_001199352.1 | NP_001186281.1 | 11 | 1031 | |||
NM_012040.3 | NP_036170.1 | 11 | 1107 | |||
GenBank mRNA | AB023027.1 | - | 10 | 977 | ||
AF181984.1 | - | 11 | 1081 | |||
AK134665.1 | - | 11 | 1038 | |||
AK158755.1 | - | 11 | 1032 | |||
BC051996.1 | - | 11 | 1014 | |||
BC055891.1 | - | 11 | 1007 | 4208888 | ||
CD776443.1 | - | 2 | 472 |