Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040682.1 | NP_001035772.1 | 9 | 1496 | ||
NM_053155.2 | NP_444385.2 | 9 | 1496 | |||
GenBank mRNA | AB047978.1 | - | 9 | 1489 | ||
AB059643.1 | - | 9 | 1489 | |||
AB059644.1 | - | 9 | 1489 | |||
AB059645.1 | - | 9 | 1489 | |||
AB059646.1 | - | 9 | 1489 | |||
AB059647.1 | - | 9 | 1489 | |||
AB059648.1 | - | 9 | 1489 | |||
AK077023.1 | - | 6 | 1078 | |||
AK147777.1 | - | 9 | 1393 | |||
AK162015.1 | - | 1 | 1213 | |||
BC019176.1 | - | 9 | 1427 | |||
BC096470.1 | - | 9 | 1385 | |||
BC141280.1 | - | 9 | 1429 | 9055892 |