Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025606.3 | NP_079882.2 | 4 | 558 | ||
GenBank mRNA | AB049643.1 | - | 4 | 558 | ||
AK004034.1 | - | 4 | 549 | |||
AK009687.1 | - | 4 | 543 | |||
AK011971.1 | - | 4 | 565 | |||
AK165616.1 | - | 4 | 591 | |||
BC029009.1 | - | 3 | 2570 | |||
BC044721.1 | - | 4 | 582 | 6476049 |