Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110202.1 | NP_001103672.1 | 3 | 1466 | ||
NM_001110203.1 | NP_001103673.1 | 3 | 1466 | |||
NM_001286386.1 | NP_001273315.1 | 3 | 1466 | |||
NM_001286387.1 | NP_001273316.1 | 3 | 1466 | |||
NM_001286388.1 | NP_001273317.1 | 3 | 1466 | |||
NM_053167.3 | NP_444397.2 | 3 | 1466 | |||
GenBank mRNA | AF220039.1 | - | 1 | 74 | ||
AK028280.1 | - | 3 | 1185 | |||
AK035807.1 | - | 3 | 1184 | |||
AK048875.1 | - | 3 | 1203 | |||
AK049907.1 | - | 3 | 1180 | |||
AK129109.1 | - | 3 | 1171 | |||
AK163123.1 | - | 3 | 1203 | |||
BC052034.1 | - | 3 | 1466 | 5684877 |