Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102471.1 | NP_001095941.1 | 1 | 1379 | ||
NM_033569.3 | NP_291047.2 | 1 | 1379 | |||
GenBank mRNA | AF216961.1 | - | 1 | 691 | ||
AK032006.1 | - | 1 | 1339 | |||
AK044400.1 | - | 1 | 1383 | |||
AK053711.1 | - | 1 | 1397 | |||
AK159616.1 | - | 1 | 1385 | |||
BC012956.1 | - | 1 | 1375 | |||
BC027387.1 | - | 1 | 1176 | |||
BC125033.1 | - | 1 | 691 |
Gene Symbol: Gm10199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK083199.1 | - | 1 | 370 | ||
AK158670.1 | - | 1 | 473 |