Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053191.2 | NP_444421.2 | 5, 6 | 808 | ||
GenBank mRNA | AF329196.1 | - | 5, 6 | 728 | ||
AK030960.1 | - | 5, 6 | 810 | |||
AK040961.1 | - | 5, 6 | 807 | |||
AK165584.1 | - | 4, 5 | 484 | |||
BC116705.1 | - | 5, 6 | 704 | 40102877 |