Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033573.2 | NP_291051.1 | 5 | 1432 | ||
GenBank mRNA | AF273750.1 | - | 5 | 1453 | ||
AK078394.1 | - | 5 | 1452 | |||
BC027831.1 | - | 5 | 1465 | |||
BC052072.1 | - | 5 | 1432 | 5717482 | ||
BC082784.1 | - | 5 | 1450 | 4191155 |