Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133787.2 | NP_598548.1 | 6 | 517 | ||
GenBank mRNA | AK088667.1 | - | 6 | 530 | ||
AK143289.1 | - | 6 | 483 | |||
AK145248.1 | - | 6 | 539 | |||
AK166283.1 | - | 6 | 485 | |||
BC003842.1 | - | 6 | 517 | 3495021 |