Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168270.1 | NP_001161742.1 | 15 | 1502 | ||
NM_133794.2 | NP_598555.2 | 19 | 1848 | |||
GenBank mRNA | AK003794.2 | - | 4 | 346 | ||
AK004814.1 | - | 19 | 1782 | |||
AK030554.1 | - | 19 | 1795 | |||
AK088049.1 | - | 19 | 1795 | |||
AK145632.1 | - | 19 | 1799 | |||
AK155759.1 | - | 19 | 1797 | |||
AK165454.1 | - | 19 | 1795 | |||
AK167782.1 | - | 19 | 1795 | |||
AK170826.1 | - | 19 | 1797 | |||
AK171293.1 | - | 19 | 1797 | |||
AK189159.1 | - | 72 | ||||
AK189775.1 | - | 2 | 55 | |||
AK209871.1 | - | 1 | 13 | |||
AK215937.1 | - | 1 | 24 | |||
BC023023.1 | - | 15 | 1502 | 5362353 | ||
BC079854.1 | - | 19 | 1782 | 5717082 |