Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133798.3 | NP_598559.2 | 3 | 851 | ||
GenBank mRNA | AK016004.1 | - | 3 | 440 | ||
AK146901.1 | - | 3 | 732 | |||
AK158201.1 | - | 3 | 742 | |||
AK162206.1 | - | 2 | 619 | |||
AK164329.1 | - | 3 | 727 | |||
AK166390.1 | - | 3 | 733 | |||
AK167692.1 | - | 3 | 852 | |||
BC018508.1 | - | 2 | 394 | 3156447 |