Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145380.2 | NP_663355.1 | 4 | 428 | ||
GenBank mRNA | AK077480.1 | - | 4 | 429 | ||
AK084738.1 | - | 4 | 447 | |||
AK146222.1 | - | 4 | 460 | |||
AK168066.1 | - | 4 | 445 | |||
BC005598.1 | - | 4 | 428 | 3969335 | ||
BC103795.1 | - | 4 | 428 | 30135815 | ||
BC116787.1 | - | 4 | 425 | 40126106 | ||
BC116789.1 | - | 4 | 425 | 40126108 |