Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178884.5 | NP_849215.3 | 6 | 2620 | ||
GenBank mRNA | AK029863.1 | - | 6 | 2409 | ||
AK048144.1 | - | 6 | 1812 | |||
AK162473.1 | - | 6 | 2530 | |||
BC051643.1 | - | 4 | 1172 | |||
BC056205.1 | - | 3 | 794 |