Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133835.2 | NP_598596.2 | 6 | 835 | ||
GenBank mRNA | AK157857.1 | - | 6 | 669 | ||
AK161969.1 | - | 1 | 38 | |||
AK177482.1 | - | 1 | 18 | |||
AK186117.1 | - | 1 | 18 | |||
AK187016.1 | - | 1 | 18 | |||
AK209158.1 | - | 11 | ||||
BC021811.1 | - | 6 | 786 | 3662474 |