Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133838.4 | NP_598599.2 | 6 | 1450 | ||
GenBank mRNA | AB239915.1 | - | 6 | 1361 | ||
AB239916.1 | - | 6 | 1361 | |||
AF173639.1 | - | 6 | 1366 | |||
AK147186.1 | - | 6 | 1402 | |||
AK159235.1 | - | 6 | 1411 | |||
AK159597.1 | - | 7 | 1410 | |||
AK166101.1 | - | 6 | 1461 | |||
AK169343.1 | - | 6 | 1461 | |||
BC007480.1 | - | 6 | 1490 | 3493227 | ||
BC012272.1 | - | 6 | 1383 | 3485836 |