Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp6nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080548.1 | NP_001074017.1 | 15 | 1767 | ||
NM_181399.3 | NP_852064.2 | 14 | 1606 | |||
GenBank mRNA | AK084045.1 | - | 15 | 1592 | ||
AK129037.1 | - | 15 | 1619 | |||
AK133119.1 | - | 6 | 845 | |||
AK155073.1 | - | 15 | 1768 | |||
BC051184.1 | - | 14 | 1609 |