Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153126.2 | NP_694766.1 | 8, 9 | 1001 | ||
GenBank mRNA | AK036294.1 | - | 8, 9 | 949 | ||
AK040070.1 | - | 8, 9 | 956 | |||
AK048552.1 | - | 7, 8 | 826 | |||
AK054029.1 | - | 8, 9 | 1022 | |||
AK087445.1 | - | 8, 9 | 959 | |||
AK149825.1 | - | 8, 9 | 828 | |||
BC034516.1 | - | 8, 9 | 1001 | 4458024 |