Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145145.1 | NP_660127.1 | 4 | 340 | ||
GenBank mRNA | AK053326.1 | - | 4 | 352 | ||
AK053889.1 | - | 4 | 339 | |||
AK134770.1 | - | 4 | 384 | |||
AK153984.1 | - | 4 | 439 | |||
AY007238.1 | - | 4 | 380 | |||
AY494857.1 | - | 4 | 400 | |||
BC027325.1 | - | 4 | 340 | 3981366 |