Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133847.3 | NP_598608.2 | 18 | 2151 | ||
GenBank mRNA | AB093220.2 | - | 17 | 1895 | ||
AK048158.1 | - | 18 | 1909 | |||
AK083541.1 | - | 18 | 2151 | |||
AK087048.1 | - | 18 | 1904 | |||
AK088012.1 | - | 18 | 1907 | |||
AK162435.1 | - | 18 | 1912 | |||
AK171487.1 | - | 18 | 1909 | |||
BC006741.1 | - | 4 | 588 | |||
BC071208.1 | - | 5 | 970 | |||
BC110309.1 | - | 18 | 1907 | 5324218 | ||
BC116320.2 | - | 18 | 2103 | 40096002 |