Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133847.3 | NP_598608.2 | 6 | 799 | ||
GenBank mRNA | AB093220.2 | - | 5 | 543 | ||
AK035622.1 | - | 6 | 761 | |||
AK047497.1 | - | 6 | 554 | |||
AK048158.1 | - | 6 | 557 | |||
AK083541.1 | - | 6 | 799 | |||
AK087048.1 | - | 6 | 552 | |||
AK088012.1 | - | 6 | 555 | |||
AK162435.1 | - | 6 | 560 | |||
AK171487.1 | - | 6 | 557 | |||
AK206609.1 | - | 2 | 117 | |||
BC110309.1 | - | 6 | 555 | 5324218 | ||
BC116320.2 | - | 6 | 751 | 40096002 |