Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133847.3 | NP_598608.2 | 17 | 1998 | ||
GenBank mRNA | AB093220.2 | - | 16 | 1742 | ||
AK035622.1 | - | 17 | 1960 | |||
AK048158.1 | - | 17 | 1756 | |||
AK083541.1 | - | 17 | 1998 | |||
AK087048.1 | - | 17 | 1751 | |||
AK088012.1 | - | 17 | 1754 | |||
AK162435.1 | - | 17 | 1759 | |||
AK171487.1 | - | 17 | 1756 | |||
BC006741.1 | - | 3 | 435 | |||
BC071208.1 | - | 4 | 817 | |||
BC110309.1 | - | 17 | 1754 | 5324218 | ||
BC116320.2 | - | 17 | 1950 | 40096002 |