Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Garnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178888.4 | NP_849219.2 | 9, 10 | 724 | ||
GenBank mRNA | AK034068.1 | - | 9, 10 | 726 | ||
AK036258.1 | - | 8, 9 | 1021 | |||
AK133163.1 | - | 10, 11 | 898 | |||
BC057383.1 | - | 8, 9 | 718 | |||
BC132143.1 | - | 9, 10 | 667 | 40130420 |