Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178890.3 | NP_849221.2 | 9, 10 | 2406 | ||
GenBank mRNA | AK042322.1 | - | 9, 10 | 2406 | ||
AK151620.1 | - | 9, 10 | 2402 | |||
AK152163.1 | - | 9, 10 | 2402 | |||
AK170569.1 | - | 9, 10 | 2405 | |||
AK172414.1 | - | 9, 10 | 2404 | |||
AK172468.1 | - | 9, 10 | 2403 | |||
AK178574.1 | - | 1, 2 | 139 | |||
BC054399.1 | - | 9, 10 | 2406 | 6493840 |