Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133857.3 | NP_598618.1 | 12 | 2025 | ||
GenBank mRNA | AK036876.1 | - | 12 | 2026 | ||
BC022221.1 | - | 2 | 167 | |||
BC132339.1 | - | 11 | 1758 | 40130616 | ||
BC145707.1 | - | 11 | 1758 | 40131010 |