Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mob3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175308.4 | NP_780517.1 | 4 | 832 | ||
GenBank mRNA | AK084014.1 | - | 4 | 833 | ||
AK148476.1 | - | 4 | 833 | |||
BC099740.1 | - | 4 | 818 | |||
BC137587.1 | - | 4 | 714 | 8860606 | ||
BC137589.1 | - | 4 | 714 | 8860608 |