Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noc4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153570.2 | NP_705798.2 | 12, 13 | 1282 | ||
GenBank mRNA | AK052601.1 | - | 12, 13 | 1282 | ||
AK088355.1 | - | 12, 13 | 1280 | |||
AK090024.1 | - | 12, 13 | 1294 | |||
AK146006.1 | - | 12, 13 | 1278 | |||
AK172310.1 | - | 12, 13 | 1261 | |||
BC024616.1 | - | 12, 13 | 1272 | 4218310 |