Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133913.2 | NP_598674.2 | 2 | 1920 | ||
GenBank mRNA | AK089404.1 | - | 2 | 1930 | ||
AK122504.1 | - | 2 | 1920 | |||
AK138814.1 | - | 1926 |