Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133932.2 | NP_598693.1 | 9 | 1401 | ||
GenBank mRNA | AF383154.1 | - | 9 | 1220 | ||
AK078906.1 | - | 9 | 1721 | |||
BC022707.1 | - | 7 | 828 | 4486171 | ||
BC026630.1 | - | 9 | 1401 | 4983657 | ||
BC032195.1 | - | 10 | 1265 | 5323331 |