Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pwwp2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033206.2 | NP_001028378.1 | 2 | 596 | ||
NM_001098636.1 | NP_001092106.1 | 2 | 596 | |||
GenBank mRNA | AK158245.1 | - | 2 | 596 | ||
AK160932.1 | - | 2 | 518 | |||
AK165809.1 | - | 2 | 593 | |||
BC116669.1 | - | 2 | 596 | 40107062 | ||
BC116720.1 | - | 2 | 592 | 40107061 |