Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apof | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133997.2 | NP_598758.1 | 2 | 808 | ||
GenBank mRNA | BC010815.1 | - | 2 | 70 | 4211838 | |
BC022795.1 | - | 2 | 107 | 5102555 | ||
BC024769.1 | - | 2 | 95 | 5123598 | ||
BC025827.1 | - | 2 | 82 | 5101712 |