Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159591.1 | NP_001153063.1 | 9 | 1284 | ||
NM_134002.2 | NP_598763.1 | 8 | 1461 | |||
GenBank mRNA | AK077076.1 | - | 9 | 1264 | ||
AK132871.1 | - | 9 | 1285 | |||
AK152413.1 | - | 8 | 1460 | |||
BC004839.1 | - | 8 | 1403 | 2648652 | ||
BC063083.1 | - | 6 | 2585 |