Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134023.1 | NP_598784.1 | 7 | 793 | ||
GenBank mRNA | AK140872.1 | - | 7 | 805 | ||
AK208062.1 | - | 3 | 168 | |||
AK220243.1 | - | 1341 | ||||
BC018300.1 | - | 7 | 793 | 4193046 | ||
BC029773.1 | - | 5 | 357 | |||
BC037230.1 | - | 7 | 776 | 2650923 |