Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027874.2 | NP_082150.1 | 7 | 1254 | ||
NM_139059.2 | NP_620690.1 | 7 | 1254 | |||
GenBank mRNA | AK051052.1 | - | 7 | 1253 | ||
AK088642.1 | - | 7 | 989 | |||
AK140317.1 | - | 7 | 979 | |||
AK143534.1 | - | 7 | 955 | |||
AK146600.1 | - | 7 | 1244 | |||
AK150387.1 | - | 7 | 983 | |||
AK150815.1 | - | 7 | 981 | |||
AK152721.1 | - | 7 | 1196 | |||
AK157812.1 | - | 7 | 989 | |||
BC004604.1 | - | 7 | 1240 | 3585286 |