Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027874.2 | NP_082150.1 | 4 | 675 | ||
NM_139059.2 | NP_620690.1 | 4 | 675 | |||
GenBank mRNA | AK004606.1 | - | 4 | 435 | ||
AK051052.1 | - | 4 | 674 | |||
AK088642.1 | - | 4 | 410 | |||
AK140317.1 | - | 4 | 400 | |||
AK143534.1 | - | 4 | 375 | |||
AK146600.1 | - | 4 | 665 | |||
AK150387.1 | - | 4 | 403 | |||
AK150815.1 | - | 4 | 402 | |||
AK152721.1 | - | 4 | 617 | |||
AK157812.1 | - | 4 | 410 | |||
AK199003.1 | - | 2 | 134 | |||
AK209498.1 | - | 2 | 136 | |||
AK210923.1 | - | 3 | 263 | |||
BC004604.1 | - | 4 | 661 | 3585286 |