Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253847.1 | NP_001240776.1 | 6 | 1068 | ||
NM_134048.2 | NP_598809.1 | 6 | 1065 | |||
GenBank mRNA | AF167441.1 | - | 6 | 1068 | ||
AK037706.1 | - | 3 | 652 | |||
AK051737.1 | - | 6 | 1007 | |||
BC019529.1 | - | 6 | 989 | 4219836 |