Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134052.2 | NP_598813.1 | 3, 4 | 455 | ||
GenBank mRNA | AK089966.1 | - | 3, 4 | 439 | ||
AK145831.1 | - | 3, 4 | 432 | |||
BC005695.1 | - | 3, 4 | 455 | 3597652 |