Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028821.3 | NP_083097.2 | 6 | 364 | ||
GenBank mRNA | AK005826.1 | - | 6 | 450 | ||
BC070454.1 | - | 321 | ||||
BC125392.1 | - | 5 | 346 | 40129907 | ||
BC125394.1 | - | 5 | 342 | 40129909 | ||
BY126068.1 | - | 3 | 160 |