Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145934.2 | NP_666046.1 | 8 | 758 | ||
GenBank mRNA | AK050131.1 | - | 8 | 738 | ||
AK050172.1 | - | 8 | 726 | |||
BC026642.1 | - | 8 | 732 | 4986113 |