Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028266.5 | NP_082542.3 | 3, 4 | 416 | ||
GenBank mRNA | AK040971.1 | - | 3, 4 | 369 | ||
AK051511.1 | - | 3, 4 | 372 | |||
AK142403.1 | - | 3, 4 | 348 | |||
BC027766.1 | - | 3, 4 | 303 | 5346676 |