Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013468.3 | NP_038496.2 | 2, 3 | 262 | ||
GenBank mRNA | AF041847.1 | - | 2, 3 | 213 | ||
AK009655.1 | - | 2, 3 | 255 | |||
AK009959.1 | - | 2, 3 | 261 | |||
AK145944.1 | - | 2, 3 | 303 | |||
AK146471.1 | - | 2, 3 | 259 | |||
AK146627.1 | - | 2, 3 | 261 | |||
AK146940.1 | - | 2, 3 | 259 | |||
BC037138.1 | - | 2, 3 | 262 | 4953125 |