Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sft2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145512.4 | NP_663487.1 | 2 | 251 | ||
GenBank mRNA | AK036578.1 | - | 2 | 139 | ||
AK075812.1 | - | 2 | 167 | |||
AK156973.1 | - | 2 | 142 | |||
AK170590.1 | - | 2 | 142 | |||
BC017549.1 | - | 2 | 130 | 4924681 |