Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025350.3 | NP_079626.2 | 3 | 599 | ||
GenBank mRNA | AK003088.1 | - | 3 | 384 | ||
BC052661.1 | - | 3 | 613 | 30050094 |