Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029787.2 | NP_084063.1 | 7, 8 | 715 | ||
GenBank mRNA | AF332059.1 | - | 7, 8 | 639 | ||
AF332060.1 | - | 7, 8 | 639 | |||
AK002640.1 | - | 7, 8 | 687 | |||
AK010858.1 | - | 7, 8 | 692 | |||
AK128917.1 | - | 7, 8 | 709 | |||
AK178355.1 | - | 3, 4 | 213 | |||
AK181262.1 | - | 3, 4 | 213 | |||
AK183532.1 | - | 3, 4 | 213 | |||
AK187743.1 | - | 3, 4 | 214 | |||
AK190014.1 | - | 3, 4 | 213 | |||
AK211950.1 | - | 3, 4 | 213 | |||
AK213922.1 | - | 3, 4 | 213 | |||
BC004760.1 | - | 7, 8 | 722 | 3154531 | ||
BC032013.1 | - | 7, 8 | 697 | 4238275 | ||
BC043074.1 | - | 7, 8 | 715 | 6404746 |